COMH04236422 Combi-Blocks, Inc.

5-(4-Fluorophenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD01194649
  • Cas Number : 310451-83-3
  • Molecular Weight : 325.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6776

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6776
Availability details for
Quantity
  • Availability
  • Add to Cart