COMH04236436 Combi-Blocks, Inc.

(1E)-1-(9H-Fluoren-2-yl)ethanone oxime

  • MDL number MFCD00558974
  • Cas Number : 110827-07-1
  • Molecular Weight : 223.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6796

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6796
Availability details for
Quantity
  • Availability
  • Add to Cart