COMH04236455 Combi-Blocks, Inc.

2-Bromo-n-(4-butoxyphenyl)-2-methylpropanamide

  • MDL number MFCD22056623
  • Cas Number : 23532-45-8
  • Molecular Weight : 314.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6828

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6828
Availability details for
Quantity
  • Availability
  • Add to Cart