COMH04236459 Combi-Blocks, Inc.

5-Oxo-1-(1-phenylethyl)pyrrolidine-3-carbonyl chloride

  • MDL number MFCD11941438
  • Cas Number : 1160264-09-4
  • Molecular Weight : 251.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6832
Availability details for
Quantity
  • Availability
  • Add to Cart