COMH042364CB Combi-Blocks, Inc.

1,3-Dioxo-2-[3-(trifluoromethyl)phenyl]isoindoline-5-carboxylic acid

  • MDL number MFCD01174923
  • Cas Number : 328555-35-7
  • Molecular Weight : 335.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6946

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6946
Availability details for
Quantity
  • Availability
  • Add to Cart