COMH042364CC Combi-Blocks, Inc.

1-Benzoylpiperidine-3-carboxylic acid

  • MDL number MFCD01856779
  • Cas Number : 13850-76-5
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6947

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6947
Availability details for
Quantity
  • Availability
  • Add to Cart