COMH042364D7 Combi-Blocks, Inc.

2-[(5-([(3-Chlorophenyl)amino]methyl)-4-methyl-4h-1,2,4-triazol-3-yl)thio]acetohydrazide

  • MDL number MFCD11696434
  • Cas Number : 1071368-84-7
  • Molecular Weight : 326.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6958

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-6958
Availability details for
Quantity
  • Availability
  • Add to Cart