COMH04236534 Combi-Blocks, Inc.

2-(Chloromethyl)-1,4,5-trimethyl-1h-imidazole hydrochloride

  • MDL number MFCD18071282
  • Cas Number : 1251762-13-6
  • Molecular Weight : 195.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7051

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7051
Availability details for
Quantity
  • Availability
  • Add to Cart