COMH0423653D Combi-Blocks, Inc.

1-(2-Nitrobenzoyl)pyrrolidine

  • MDL number MFCD00594851
  • Cas Number : 169330-07-8
  • Molecular Weight : 220.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7060

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7060
Availability details for
Quantity
  • Availability
  • Add to Cart