COMH0423653F Combi-Blocks, Inc.

2-[(Cyclohexylcarbonyl)amino]benzoic acid

  • MDL number MFCD00223756
  • Cas Number : 25437-73-4
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7062

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7062
Availability details for
Quantity
  • Availability
  • Add to Cart