COMH04236556 Combi-Blocks, Inc.

4-Methyl-1h-benzimidazole-2-sulfonic acid

  • MDL number MFCD11696447
  • Cas Number : 1071368-98-3
  • Molecular Weight : 212.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7085

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7085
Availability details for
Quantity
  • Availability
  • Add to Cart