COMH04236592 Combi-Blocks, Inc.

7-Hydroxy-4-(4-methoxyphenyl)-8-methyl-2h-chromen-2-one

  • MDL number MFCD02222114
  • Cas Number : 370583-62-3
  • Molecular Weight : 282.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7145

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7145
Availability details for
Quantity
  • Availability
  • Add to Cart