COMH042365B0 Combi-Blocks, Inc.

4-Bromo-5-propylthiophene-2-carbonyl chloride

  • MDL number MFCD12197882
  • Cas Number : 1160249-08-0
  • Molecular Weight : 267.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7175

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7175
Availability details for
Quantity
  • Availability
  • Add to Cart