COMH0423661B Combi-Blocks, Inc.

1-(2-Ethoxyphenyl)-1h-pyrrole-2,5-dione

  • MDL number MFCD00030662
  • Cas Number : 36817-57-9
  • Molecular Weight : 217.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7282

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7282
Availability details for
Quantity
  • Availability
  • Add to Cart