COMH04236634 Combi-Blocks, Inc.

1-[2-(4,5-Dimethoxy-2-nitrophenyl)ethyl]pyrrolidin-2-one

  • MDL number MFCD12972456
  • Cas Number : 1171604-89-9
  • Molecular Weight : 294.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7307

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7307
Availability details for
Quantity
  • Availability
  • Add to Cart