COMH04236635 Combi-Blocks, Inc.

5-Ethyl-4,5,6,7-tetrahydro-2h-pyrazolo[4,3-c]pyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD13248699
  • Cas Number : 1216236-82-6
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7308

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7308
Availability details for
Quantity
  • Availability
  • Add to Cart