COMH042366A0 Combi-Blocks, Inc.

1-(4-Nitrophenyl)-2,3,4,9-tetrahydro-1h-beta-carboline hydrochloride

  • MDL number MFCD07626284
  • Cas Number : 3380-77-6
  • Molecular Weight : 329.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7415

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7415
Availability details for
Quantity
  • Availability
  • Add to Cart