COMH042366A5 Combi-Blocks, Inc.

5-Methoxy-1-methyl-1h-indole-2-carbohydrazide

  • MDL number MFCD00449859
  • Cas Number : 200062-20-0
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7420

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7420
Availability details for
Quantity
  • Availability
  • Add to Cart