COMH042366C3 Combi-Blocks, Inc.

4-[(2,6-Dichlorobenzyl)oxy]-3-ethoxybenzoic acid

  • MDL number MFCD12028126
  • Cas Number : 1142201-94-2
  • Molecular Weight : 341.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7450

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7450
Availability details for
Quantity
  • Availability
  • Add to Cart