COMH042366D2 Combi-Blocks, Inc.

4-Methyl-3-[(4-methylpiperazin-1-yl)sulfonyl]benzoic acid

  • MDL number MFCD13353554
  • Cas Number : 1036541-46-4
  • Molecular Weight : 298.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7465

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7465
Availability details for
Quantity
  • Availability
  • Add to Cart