COMH042366F6 Combi-Blocks, Inc.

3-Methoxy-4-[(4-methylbenzyl)oxy]benzoyl chloride

  • MDL number MFCD12197953
  • Cas Number : 1160250-76-9
  • Molecular Weight : 290.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7501

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7501
Availability details for
Quantity
  • Availability
  • Add to Cart