COMH042366F8 Combi-Blocks, Inc.

5,6-Dimethoxy-1h-indole-2-carbohydrazide

  • MDL number MFCD07364708
  • Cas Number : 121282-81-3
  • Molecular Weight : 235.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7503

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7503
Availability details for
Quantity
  • Availability
  • Add to Cart