COMH0423671E Combi-Blocks, Inc.

(1,1-Dimethyl-2-oxo-2-piperidin-1-ylethyl)amine hydrochloride

  • MDL number MFCD13562095
  • Cas Number : 1220034-81-0
  • Molecular Weight : 206.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7541

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7541
Availability details for
Quantity
  • Availability
  • Add to Cart