COMH0423673C Combi-Blocks, Inc.

2-Cyano-n-cyclopropylacetamide

  • MDL number MFCD02176352
  • Cas Number : 15029-37-5
  • Molecular Weight : 124.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7571

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7571
Availability details for
Quantity
  • Availability
  • Add to Cart