COMH04236744 Combi-Blocks, Inc.

4-Amino-n-(2,5-dimethylphenyl)benzenesulfonamide

  • MDL number MFCD01016107
  • Cas Number : 188357-46-2
  • Molecular Weight : 276.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7579
Availability details for
Quantity
  • Availability
  • Add to Cart