COMH04236755 Combi-Blocks, Inc.

2-[2-(Methylsulfonyl)ethyl]piperidine hydrochloride

  • MDL number MFCD26793023
  • Cas Number : 1803585-98-9
  • Molecular Weight : 227.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7596
Availability details for
Quantity
  • Availability
  • Add to Cart