COMH0423677F Combi-Blocks, Inc.

3-(4-Methyl-1h-indol-1-yl)propanoic acid

  • MDL number MFCD11212266
  • Cas Number : 1094654-47-3
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7638

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7638
Availability details for
Quantity
  • Availability
  • Add to Cart