COMH0423679C Combi-Blocks, Inc.

4-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]-4-oxobutanoic acid

  • MDL number MFCD10007609
  • Cas Number : 1017384-80-3
  • Molecular Weight : 320.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7667

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7667
Availability details for
Quantity
  • Availability
  • Add to Cart