COMH0423679F Combi-Blocks, Inc.

N'-(1,1-Dioxido-1,2-benzisothiazol-3-yl)-4-methylbenzenesulfonohydrazide

  • MDL number MFCD00760675
  • Cas Number : 13255-93-1
  • Molecular Weight : 351.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7670

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7670
Availability details for
Quantity
  • Availability
  • Add to Cart