COMH042367F2 Combi-Blocks, Inc.

(1-[2-(4-Methylphenoxy)ethyl]-1h-benzimidazol-2-yl)methanol

  • MDL number MFCD01141653
  • Cas Number : 313067-45-7
  • Molecular Weight : 282.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7753
Availability details for
Quantity
  • Availability
  • Add to Cart