COMH04236800 Combi-Blocks, Inc.

N-(2,3-Dichlorophenyl)imidodicarbonimidic diamide

  • MDL number MFCD00276882
  • Cas Number : 143413-10-9
  • Molecular Weight : 246.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7767

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7767
Availability details for
Quantity
  • Availability
  • Add to Cart