COMH04236851 Combi-Blocks, Inc.

2-[1-(1,1-Dioxido-1,2-benzisothiazol-3-yl)hydrazino]ethanol

  • MDL number MFCD01446002
  • Cas Number : 312500-56-4
  • Molecular Weight : 241.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7848

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7848
Availability details for
Quantity
  • Availability
  • Add to Cart