COMH042368AD Combi-Blocks, Inc.

2-(3-Butoxyphenyl)-8-methylquinoline-4-carbonyl chloride

  • MDL number MFCD03421220
  • Cas Number : 1160254-67-0
  • Molecular Weight : 353.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7940

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7940
Availability details for
Quantity
  • Availability
  • Add to Cart