COMH042368B1 Combi-Blocks, Inc.

1,5-Dimethyl-1h-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD01114739
  • Cas Number : 329064-07-5
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7944

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7944
Availability details for
Quantity
  • Availability
  • Add to Cart