COMH042368E4 Combi-Blocks, Inc.

2-(4-Benzyl-5-mercapto-4h-1,2,4-triazol-3-yl)phenol

  • MDL number MFCD01439946
  • Cas Number : 312319-66-7
  • Molecular Weight : 283.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7995

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-7995
Availability details for
Quantity
  • Availability
  • Add to Cart