COMH04236931 Combi-Blocks, Inc.

[(5-Methyl-4-phenyl-4h-1,2,4-triazol-3-yl)thio]acetic acid

  • MDL number MFCD02227063
  • Cas Number : 200815-83-4
  • Molecular Weight : 249.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8072

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8072
Availability details for
Quantity
  • Availability
  • Add to Cart