COMH0423695C Combi-Blocks, Inc.

N-[2-(Dimethylamino)ethyl]-2-(methylamino)acetamide

  • MDL number MFCD09943853
  • Cas Number : 1016800-40-0
  • Molecular Weight : 159.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8115
Availability details for
Quantity
  • Availability
  • Add to Cart