COMH04236971 Combi-Blocks, Inc.

[5-Ethyl-3-(morpholin-4-ylcarbonyl)-2-thienyl]amine

  • MDL number MFCD11653238
  • Cas Number : 1099685-01-4
  • Molecular Weight : 240.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8136
Availability details for
Quantity
  • Availability
  • Add to Cart