COMH04236983 Combi-Blocks, Inc.

2-(3-Bromophenyl)-8-chloroquinoline-4-carbonyl chloride

  • MDL number MFCD03422827
  • Cas Number : 1160263-63-7
  • Molecular Weight : 381.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8154

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8154
Availability details for
Quantity
  • Availability
  • Add to Cart