COMH0423698E Combi-Blocks, Inc.

3-[4-(4-Hydroxyphenoxy)-3,5-diiodophenyl]propanoic acid

  • MDL number MFCD00045945
  • Cas Number : 1158-10-7
  • Molecular Weight : 510.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8165

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8165
Availability details for
Quantity
  • Availability
  • Add to Cart