COMH04236A9A Combi-Blocks, Inc.

1-Benzylcyclohexanol

  • MDL number MFCD00155242
  • Cas Number : 1944-01-0
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8434

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8434
Availability details for
Quantity
  • Availability
  • Add to Cart