COMH04236A9F Combi-Blocks, Inc.

1-[3-(Methylamino)propyl]-1,3-dihydro-2h-benzimidazol-2-one hydrochloride

  • MDL number MFCD11844251
  • Cas Number : 1172831-89-8
  • Molecular Weight : 241.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8439

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8439
Availability details for
Quantity
  • Availability
  • Add to Cart