COMH04236AAD Combi-Blocks, Inc.

tert-Butyl [1-(2-hydrazino-2-oxoethyl)piperidin-4-yl]carbamate

  • MDL number MFCD29035066
  • Cas Number : 1463524-40-4
  • Molecular Weight : 272.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8453

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8453
Availability details for
Quantity
  • Availability
  • Add to Cart