COMH04236AB0 Combi-Blocks, Inc.

3-(3-Ethyl-1-phenyl-1h-1,2,4-triazol-5-yl)propanoic acid

  • MDL number MFCD12028273
  • Cas Number : 1142209-71-9
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8456

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8456
Availability details for
Quantity
  • Availability
  • Add to Cart