COMH04236AE5 Combi-Blocks, Inc.

Ethyl 3-(4-chlorophenyl)-1,2,4-oxadiazole-5-carboxylate

  • MDL number MFCD05667194
  • Cas Number : 163719-69-5
  • Molecular Weight : 252.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8509

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8509
Availability details for
Quantity
  • Availability
  • Add to Cart