COMH04236B02 Combi-Blocks, Inc.

Ethyl (4-methyl-4-phenyl-3,4-dihydroquinolin-1(2h)-yl)(oxo)acetate

  • MDL number MFCD19103601
  • Cas Number : 1306738-90-8
  • Molecular Weight : 323.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8538

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8538
Availability details for
Quantity
  • Availability
  • Add to Cart