COMH04236B80 Combi-Blocks, Inc.

4-Chloro-2-(5-mercapto-4-methyl-4h-1,2,4-triazol-3-yl)phenol

  • MDL number MFCD01995824
  • Cas Number : 273216-76-5
  • Molecular Weight : 241.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8664

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8664
Availability details for
Quantity
  • Availability
  • Add to Cart