COMH04236B87 Combi-Blocks, Inc.

4-(4-Bromo-5-methyl-3-nitro-1h-pyrazol-1-yl)butanoic acid

  • MDL number MFCD12028393
  • Cas Number : 1171922-90-9
  • Molecular Weight : 292.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8671

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8671
Availability details for
Quantity
  • Availability
  • Add to Cart