COMH04236B8A Combi-Blocks, Inc.

6-Methyl-2-pyridin-3-ylquinoline-4-carbonyl chloride hydrochloride

  • MDL number MFCD18205925
  • Cas Number : 1203205-47-3
  • Molecular Weight : 319.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8674

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8674
Availability details for
Quantity
  • Availability
  • Add to Cart