COMH04236B94 Combi-Blocks, Inc.

4-(5-tert-Butyl-1,2,4-oxadiazol-3-yl)benzoic acid

  • MDL number MFCD12026875
  • Cas Number : 1119452-72-0
  • Molecular Weight : 246.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8684

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-8684
Availability details for
Quantity
  • Availability
  • Add to Cart